Tris(3-methoxyphenyl)phosphine, 98%
Tris(3-methoxyphenyl)phosphine, 98%
Tris(3-methoxyphenyl)phosphine, 98%
Tris(3-methoxyphenyl)phosphine, 98%
Thermo Scientific Chemicals

Tris(3-methoxyphenyl)phosphine, 98%

CAS: 29949-84-6 | C21H21O3P | 352.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A17949.06
also known as A17949-06
Price (USD)
249.65
Special offer
Online exclusive
Ends: 30-Jun-2026
294.00
Save 44.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
249.65
Special offer
Online exclusive
Ends: 30-Jun-2026
294.00
Save 44.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS29949-84-6
IUPAC Nametris(3-methoxyphenyl)phosphane
Molecular FormulaC21H21O3P
InChI KeyCCXTYQMZVYIQRP-UHFFFAOYSA-N
SMILESCOC1=CC(=CC=C1)P(C1=CC(OC)=CC=C1)C1=CC=CC(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)111.0-118.0?C
Appearance (Color)White to cream
Purity (DSC)≥97.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only