4-Hydroxy-3-nitrobenzaldehyde, 98%
4-Hydroxy-3-nitrobenzaldehyde, 98%
4-Hydroxy-3-nitrobenzaldehyde, 98%
4-Hydroxy-3-nitrobenzaldehyde, 98%
Thermo Scientific Chemicals

4-Hydroxy-3-nitrobenzaldehyde, 98%

CAS: 3011-34-5 | C7H5NO4 | 167.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17972.14
also known as A17972-14
Price (USD)
234.65
Special offer
Online exclusive
Ends: 30-Jun-2026
276.00
Save 41.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
234.65
Special offer
Online exclusive
Ends: 30-Jun-2026
276.00
Save 41.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS3011-34-5
IUPAC Name4-hydroxy-3-nitrobenzaldehyde
Molecular FormulaC7H5NO4
InChI KeyYTHJCZRFJGXPTL-UHFFFAOYSA-N
SMILESOC1=CC=C(C=O)C=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Pale yellow to yellow to brown
Assay (GC)≥97.5%
Melting Point (clear melt)137.0-145.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only