1-Butylphosphonic acid, 98%
1-Butylphosphonic acid, 98%
1-Butylphosphonic acid, 98%
Thermo Scientific Chemicals

1-Butylphosphonic acid, 98%

CAS: 3321-64-0 | C4H11O3P | 138.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A17983.03
also known as A17983-03
Price (USD)
54.20
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
54.20
Each
Chemical Identifiers
CAS3321-64-0
IUPAC Namebutylphosphonic acid
Molecular FormulaC4H11O3P
InChI KeyUOKRBSXOBUKDGE-UHFFFAOYSA-N
SMILESCCCCP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or flakes
Assay (Aqueous acid-base Titration)>97.5 to <102.5%
Water Content (Karl Fischer Titration)<2.0%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only