(-)-Diethyl D-tartrate, 99%
(-)-Diethyl D-tartrate, 99%
(-)-Diethyl D-tartrate, 99%
Thermo Scientific Chemicals

(-)-Diethyl D-tartrate, 99%

CAS: 13811-71-7 | C8H14O6 | 206.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A17992.22
also known as A17992-22
Price (USD)
199.65
Online Exclusive
222.00
Save 22.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
199.65
Online Exclusive
222.00
Save 22.35 (10%)
Each
Chemical Identifiers
CAS13811-71-7
IUPAC Name1,4-diethyl (2S,3S)-2,3-dihydroxybutanedioate
Molecular FormulaC8H14O6
InChI KeyYSAVZVORKRDODB-WDSKDSINSA-N
SMILESCCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid or viscous liquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Optical Rotation-7.5° to -9.5° (neat)
View more
Diethyl D(-)-tartrate is mainly used in chiral pharmaceuticals, chiral intermediates and chiral catalysts. It can be used with titanium in enolic asymmetric epoxidization. The compound d-alpha-tocopherol is the principal compound present in natural vitamin E sources. At present, d-alpha-tocopherol and other tocopherol derivatives are used in pharmaceuticals, foods and animal feeds. It is used as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl D(-)-tartrate is mainly used in chiral pharmaceuticals, chiral intermediates and chiral catalysts. It can be used with titanium in enolic asymmetric epoxidization. The compound d-alpha-tocopherol is the principal compound present in natural vitamin E sources. At present, d-alpha-tocopherol and other tocopherol derivatives are used in pharmaceuticals, foods and animal feeds. It is used as a food additive.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Jiang Weng.; Yong-Bo Li.; Rui-Bin Wang.; Feng-Quan Li.; Can Liu.; Albert S. C. Chan.; and Gui Lu. A Practical and Azide-Free Synthetic Approach to Oseltamivir from Diethyl d-Tartrate.J. Org. Chem.,2010,75(9), 3125-3128.
  2. Simon Azeroual.; Jamie Surprenant.; Thomas D. Lazzara.; Marta Kocun.; Ye Tao.; Louis A. Cuccia.; and Jean-Marie Lehn. Mirror symmetry breaking and chiral amplification in foldamer-based supramolecular helical aggregates.Chem. Commun.,2012,48(17), 2292-2294.