It is employed in the spectrophotometric analysis determination of methenamine and its salts. It is also used in the detection and repair mechanism of Etheno-DNA adducts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the spectrophotometric analysis determination of methenamine and its salts. It is also used in the detection and repair mechanism of Etheno-DNA adducts.
Solubility
Solubility in hot methanol almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
- Shivange G.; Kodipelli N.; Anindya R. 2-Hydrazinobenzothiazole-based etheno-adduct repair protocol (HERP): a method for quantitative determination of direct repair of etheno-bases.DNA Repair (Amst).,2015,288-13.
- Lim JK.; Chen CCC. Spectrophotometric determination of methenamine and its salts by 2-hydrazinobenzothiazole.J. Pharm. Sci..,1973,621503-1504.