5-Sulfoisophthalic acid monosodium salt, 95%
5-Sulfoisophthalic acid monosodium salt, 95%
5-Sulfoisophthalic acid monosodium salt, 95%
Thermo Scientific Chemicals

5-Sulfoisophthalic acid monosodium salt, 95%

CAS: 6362-79-4 | C8H5NaO7S | 268.171 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
500 g
Catalog number A18028.30
also known as A18028-30
Price (USD)
74.65
Online Exclusive
83.20
Save 8.55 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
74.65
Online Exclusive
83.20
Save 8.55 (10%)
Each
Chemical Identifiers
CAS6362-79-4
IUPAC Namesodium 3,5-dicarboxybenzene-1-sulfonate
Molecular FormulaC8H5NaO7S
InChI KeyYXTFRJVQOWZDPP-UHFFFAOYSA-M
SMILES[Na+].OC(=O)C1=CC(=CC(=C1)C(O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
It can be used as the third monomer in the Polyester procession of esterification, which can improve the ability of product's dying. And it can be used as the catalyst in medicine, agricultural chemical, polyester. Used as a monomer for waterborne polyesters and nylon fibers, in adhesion aid for magnetic tapes, and amine free polyurethane dispersions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as the third monomer in the Polyester procession of esterification, which can improve the ability of product′s dying. And it can be used as the catalyst in medicine, agricultural chemical, polyester. Used as a monomer for waterborne polyesters and nylon fibers, in adhesion aid for magnetic tapes, and amine free polyurethane dispersions.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Qing-Yan Liu; and Li Xu. Synthesis, Crystal Structures, and Photophysical Properties of Two Novel Lead(II)-SIP Coordination Polymers (NaH2SIP = 5-Sulfoisophthalic Acid Monosodium Salt) Containing Tetranuclear Lead(II) Units. European Journal of Inorganic Chemistry. 2006, 2006 (8), 1620-1628.
  2. Yu-Ling Wang; Qing-Yan Liu and Li Xu. Two novel luminescent silver(I) coordination polymers containing octanuclear silver cluster units or ligand unsupported Ag⋯Ag interactions constructed from 5-sulfoisophthalic acid (H3SIP) and organic amine.CrystEngComm. 2008, 10 (11), 1667-1673.