Phosphoric acid, 85% aq. soln.
Phosphoric acid, 85% aq. soln.
Phosphoric acid, 85% aq. soln.
Phosphoric acid, 85% aq. soln.
Thermo Scientific Chemicals

Phosphoric acid, 85% aq. soln.

CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
500 mL
2500 mL
Catalog number A18067.AP
Price (USD)
45.65
Online Exclusive
50.40
Save 4.75 (9%)
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
45.65
Online Exclusive
50.40
Save 4.75 (9%)
Each
Add to cart
Chemical Identifiers
CAS7664-38-2
IUPAC Namephosphoric acid
Molecular FormulaH3O4P
InChI KeyNBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILESOP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormViscous liquid or low-melting solid
Assay (Aqueous acid-base Titration)84.0-87.0% (non-U.S. specification)
Assay from Suppliers CofA84.0-87.0% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
Phosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It is also used in dentistry and orthodontics as an etching solution as well as a sanitizing agent in dairy, food and brewing industries. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion in to water soluble phosphate compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It is also used in dentistry and orthodontics as an etching solution as well as a sanitizing agent in dairy, food and brewing industries. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion in to water soluble phosphate compound.

Notes
Incompatible with strong bases and powdered metals.
RUO – Research Use Only

General References:

  1. Aili, D.; Vassiliev, A.; Jensen, J. O.; Schmidt, T. J.; Li, Q. Methyl phosphate formation as a major degradation mode of direct methanol fuel cells with phosphoric acid based electrolytes. J. Power Sources 2015, 279, 517-521.
  2. Bondesson, P. M.; Dupuy, A.; Galbe, M.; Zacchi, G. Optimizing Ethanol and Methane Production from Steam-pretreated, Phosphoric Acid-impregnated Corn Stover. Appl. Biochem. Biotechnol. 2015, 175 (3), 1371-1388.