2-Methoxyphenyl isocyanate, 98%
2-Methoxyphenyl isocyanate, 98%
2-Methoxyphenyl isocyanate, 98%
2-Methoxyphenyl isocyanate, 98%
Thermo Scientific Chemicals

2-Methoxyphenyl isocyanate, 98%

CAS: 700-87-8 | C8H7NO2 | 149.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18075.06
also known as A18075-06
Price (USD)
38.65
Online Exclusive
42.60
Save 3.95 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
38.65
Online Exclusive
42.60
Save 3.95 (9%)
Each
Add to cart
Chemical Identifiers
CAS700-87-8
IUPAC Name1-isocyanato-2-methoxybenzene
Molecular FormulaC8H7NO2
InChI KeySUVCZZADQDCIEQ-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.5395-1.5435 @ 20?C
Assay (GC)≥97.5%
2-Methoxyphenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxyphenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive. Protect from water and humidity. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store under dry inert gas.
RUO – Research Use Only
Jayashree Yenagi; Anita R Nandurkar; J Tonannavar. 2-Methoxyphenyl isocyanate and 2-Methoxyphenyl isothiocyanate: conformers, vibration structure and multiplet Fermi resonance. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy.2012, 91, 261-268.