trans-2-Nonenal, 97%
trans-2-Nonenal, 97%
trans-2-Nonenal, 97%
trans-2-Nonenal, 97%
Thermo Scientific Chemicals

trans-2-Nonenal, 97%

CAS: 18829-56-6 | C9H16O | 140.226 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18108.06
also known as A18108-06
Price (USD)
44.65
Special offer
Online exclusive
Ends: 31-Dec-2025
52.20
Save 7.55 (14%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 31-Dec-2025
52.20
Save 7.55 (14%)
Each
Add to cart
Chemical Identifiers
CAS18829-56-6
IUPAC Name(2E)-non-2-enal
Molecular FormulaC9H16O
InChI KeyBSAIUMLZVGUGKX-BQYQJAHWSA-N
SMILESCCCCCC\C=C\C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Free acid (titration)≤1.5%
Identification (FTIR)Conforms
View more
trans-2-Nonenal reacts with acetic acid hydrazide to produce acetic acid non-2-enylidene-hydrazide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-2-Nonenal reacts with acetic acid hydrazide to produce acetic acid non-2-enylidene-hydrazide.

Solubility
Soluble in Alcohol , and oils. Insoluble in water.

Notes
Air Sensitive. Incompatible with air, oxidizing agents and heat. Protect from heat.
RUO – Research Use Only
T Galliard; D R Phillips; J Reynolds. The formation of cis-3-nonenal, trans-2-nonenal and hexanal from linoleic acid hydroperoxide isomers by a hydroperoxide cleavage enzyme system in cucumber (Cucumis sativus) fruits. Biochimica et Biophysica Acta.1976, 441, (2), 181-192.