2-Bromobenzylamine hydrochloride, 97%
2-Bromobenzylamine hydrochloride, 97%
2-Bromobenzylamine hydrochloride, 97%
Thermo Scientific Chemicals

2-Bromobenzylamine hydrochloride, 97%

CAS: 5465-63-4 | C7H8BrClN | 221.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18151.06
also known as A18151-06
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.20
Save 11.55 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.20
Save 11.55 (14%)
Each
Chemical Identifiers
CAS5465-63-4
IUPAC Name1-(2-bromophenyl)methanamine hydrochloridyl
Molecular FormulaC7H8BrClN
InChI KeyZANQNQMPTMEAIP-UHFFFAOYSA-N
SMILES[Cl].NCC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration ex Chloride)≥96.0%
FormPowder
Melting Point (clear melt)233.5-239.5?C
It is employed as an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate in organic syntheses.

Solubility
Soluble in methanol (25 mg/ml).

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Hilton ST, et al. A tandem radical approach to the ABCE-rings of the Aspidosperma and Strychnos alkaloids.Chem. Commun. (Camb.) 2,2001,209-210.