3-Aminobenzeneboronic acid monohydrate, 97%
3-Aminobenzeneboronic acid monohydrate, 97%
3-Aminobenzeneboronic acid monohydrate, 97%
Thermo Scientific Chemicals

3-Aminobenzeneboronic acid monohydrate, 97%

CAS: 206658-89-1 | C6H10BNO3 | 154.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A18189.06
also known as A18189-06
Price (USD)
180.65
Online Exclusive
201.00
Save 20.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
180.65
Online Exclusive
201.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS206658-89-1
IUPAC Name(3-aminophenyl)boronic acid hydrate
Molecular FormulaC6H10BNO3
InChI KeyXAEOVQODHLLNKX-UHFFFAOYSA-N
SMILESO.NC1=CC=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Proton NMRConforms to structure
Appearance (Color)Pale brown
Assay (Aqueous acid-base Titration)≥96.0%
Assay (HPLC)≥96.0%
It is employed as a reagent in the preparation of Suzuki-Miyaura cross-coupling, used for Gram-positive ant virulence drugs and inhibitors of Streptococcus agalactiae Stk1, regioisomer of Zaleplon (a sedative), amphiphilic random glycopolymer, which self-assemble to form nanoparticles, with potential as a glucose-sensitive matrix, chemomechanical polymer that expands and contracts in blood plasma with high glucose selectivity. Aminophenylboronic Acid is also used as a reagent in the preparation of nonbenzodiazepine hypnotic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reagent in the preparation of Suzuki-Miyaura cross-coupling, used for Gram-positive ant virulence drugs and inhibitors of Streptococcus agalactiae Stk1, regioisomer of Zaleplon (a sedative), amphiphilic random glycopolymer, which self-assemble to form nanoparticles, with potential as a glucose-sensitive matrix, chemomechanical polymer that expands and contracts in blood plasma with high glucose selectivity. Aminophenylboronic Acid is also used as a reagent in the preparation of nonbenzodiazepine hypnotic agents.

Solubility
Soluble in DMSO and methanol.

Notes
Air & Moisture Sensitive. Store away from oxidizing agents, air, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Gregory D. Cuny.; Maxime Robin.; Natalia P. Ulyanova.; Debasis Patnaik.; Valerie Pique.; Gilles Casano.; Ji-Feng Liu.; Xiangjie Lin.; Jun Xian.; Marcie A. Glicksman.; Ross L. Stein.; Jonathan M.G. Higgins. Structure-activity relationship study of acridine analogs as haspin and DYRK2 kinase inhibitors.Bioorg Med Chem Lett.2010,20(12), 3491-3494.
  2. Xingju Jin.; Xinge Zhang.; Zhongming Wu.; Dayong Teng.; Xuejiao Zhang.; Yanxia Wang.; Zhen Wang.; Chaoxing Li. Amphiphilic random glycopolymer based on phenylboronic acid: synthesis, characterization, and potential as glucose-sensitive matrix.Biomacromolecules.2009,10(6), 1337-1345.