Isonicotinamide, 99%
Isonicotinamide, 99%
Isonicotinamide, 99%
Isonicotinamide, 99%
Thermo Scientific Chemicals

Isonicotinamide, 99%

CAS: 1453-82-3 | C6H6N2O | 122.127 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
250 g
500 g
Catalog number A18190.18
also known as A18190-18
Price (USD)
36.65
Online Exclusive
40.60
Save 3.95 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
36.65
Online Exclusive
40.60
Save 3.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS1453-82-3
IUPAC Namepyridine-4-carboxamide
Molecular FormulaC6H6N2O
InChI KeyVFQXVTODMYMSMJ-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)153.0-160.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only