1-Nonylamine, 97%
1-Nonylamine, 97%
1-Nonylamine, 97%
Thermo Scientific Chemicals

1-Nonylamine, 97%

CAS: 112-20-9 | C9H22ClN | 179.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A18209.14
also known as A18209-14
Price (USD)
105.65
Special offer
Online exclusive
Ends: 30-Jun-2026
124.00
Save 18.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
105.65
Special offer
Online exclusive
Ends: 30-Jun-2026
124.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS112-20-9
IUPAC Namenonan-1-aminium chloride
Molecular FormulaC9H22ClN
InChI KeyIMGHCWMUWVHYCO-UHFFFAOYSA-N
SMILES[Cl-].CCCCCCCCC[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4315-1.4365 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
1-Nonylamine is used as pharmaceutical intermediate. It is also used as a building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Nonylamine is used as pharmaceutical intermediate. It is also used as a building block.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
RUO – Research Use Only

General References:

  1. Lukas Meier; Stefan Schmid; Christian Berchtold; Renato Zenobi. Contribution of liquid-phase and gas-phase ionization in extractive electrospray ionization mass spectrometry of primary amines. European Journal of Mass Spectrometry. 2011, 17,(4), 345-351.
  2. T.N.Guru Row. Hydrogen and fluorine in crystal engineering: systematics from crystallographic studies of hydrogen bonded tartrate-amine complexes and fluoro-substituted coumarins, styrylcoumarins and butadienes. Coordination Chemistry Reviews. 1999, 183,(1), 81-100.