3-(4-Methoxybenzoyl)propionic acid, 98+%
3-(4-Methoxybenzoyl)propionic acid, 98+%
3-(4-Methoxybenzoyl)propionic acid, 98+%
Thermo Scientific Chemicals

3-(4-Methoxybenzoyl)propionic acid, 98+%

CAS: 3153-44-4 | C11H12O4 | 208.213 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18210.14
also known as A18210-14
Price (USD)
117.65
Special offer
Online exclusive
Ends: 28-Dec-2026
138.00
Save 20.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
117.65
Special offer
Online exclusive
Ends: 28-Dec-2026
138.00
Save 20.35
Each
Chemical Identifiers
CAS3153-44-4
IUPAC Name4-(4-methoxyphenyl)-4-oxobutanoic acid
Molecular FormulaC11H12O4
InChI KeyOMTDIBZSUZNVJK-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)144.0-150.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only