5-Bromo-2,4-dihydroxybenzoic acid, 97%
5-Bromo-2,4-dihydroxybenzoic acid, 97%
5-Bromo-2,4-dihydroxybenzoic acid, 97%
5-Bromo-2,4-dihydroxybenzoic acid, 97%
Thermo Scientific Chemicals

5-Bromo-2,4-dihydroxybenzoic acid, 97%

CAS: 7355-22-8 | C7H5BrO4 | 233.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18230.06
also known as A18230-06
Price (USD)
38.65
Online Exclusive
42.30
Save 3.65 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
38.65
Online Exclusive
42.30
Save 3.65 (9%)
Each
Add to cart
Chemical Identifiers
CAS7355-22-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0% to ≤104.0%
Assay (Silylated GC)≥96.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only