1-Hexanol, 99%
1-Hexanol, 99%
1-Hexanol, 99%
Thermo Scientific Chemicals

1-Hexanol, 99%

CAS: 111-27-3 | C6H14O | 102.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number A18232.AP
also known as A18232-AP
Price (USD)
35.70
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
35.70
Each
Chemical Identifiers
CAS111-27-3
IUPAC Namehexan-1-ol
Molecular FormulaC6H14O
InChI KeyZSIAUFGUXNUGDI-UHFFFAOYSA-N
SMILESCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms (UK Sourced Material)
CommentMaterial Sourced in UK and US
Assay from Supplier's CofA≥98.5% (US Sourced Material)
Refractive Index1.4165-1.4195 @ 20?C (UK Sourced Material)
FormLiquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1-Hexanol is used as a precursor to plasticizers, chemical intermediate for pharmaceuticals, perfume esters and antiseptics. Further, it serves as a perturbing agent on actomyosin adenosine triphosphatease. In addition to this, it is used to modulate the function of actomyosin motor.

Solubility
Miscible with ethanol, acetone, chloroform, ether, benzene. Slightly miscible with carbon tetrachloride and water.

Notes
Incompatible with strong acids, acid chlorides, acid anhydrides and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Semenov, K. N.; Regueira, T.; Fernández, J.; Charykov, N. A.; Murin, I. V. Pressure dependence of the solubility of light fullerenes in 1-hexanol from 298.15 K to 363.15 K. J. Mol. Liq. 2015, 209, 71-76.
  2. Patel, V.; Ray, D.; Singh, K.; Abezgauz, L.; Marangoni, G.; Aswal, V. K.; Bahadur, P. 1-Hexanol triggered structural characterization of the worm-like micelle to vesicle transitions in cetyltrimethylammonium tosylate solutions. RSC Adv. 2015, 5 (107), 87758-87768.