Indium(III) nitrate hydrate, In 29% min.
Indium(III) nitrate hydrate, In 29% min.
Indium(III) nitrate hydrate, In 29% min.
Thermo Scientific Chemicals

Indium(III) nitrate hydrate, In 29% min.

CAS: 207398-97-8 | InN3O9 | 300.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A18252.14
also known as A18252-14
Price (USD)
125.65
Online exclusive
139.00
Save 13.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
125.65
Online exclusive
139.00
Save 13.35
Each
Chemical Identifiers
CAS207398-97-8
IUPAC Nameindium(3+) trinitrate
Molecular FormulaInN3O9
InChI KeyLKRFCKCBYVZXTC-UHFFFAOYSA-N
SMILES[In+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused/lumpy solid
Elemental AnalysisIn: ≥29.0%
Indium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.

Solubility
Soluble in water, methanol and ethanol.

Notes
It is stable at room temperature. Hygroscopic. Incompatible with strong reducing agents and strong acids.
RUO – Research Use Only

General References:

  1. Selvakumar, D.; Dharmaraj, N.; Kumar, N. S.; Padaki, V. C. Preparation, Surface Morphology, and Optical Properties Of Indium(III) Oxide Nanoparticles by Thermolysis Of Indium-Poly(Vinyl Alcohol) Coordination Polymer. J. Coord. Chem. 2014, 67 (11), 1938-1947.
  2. Samanamú, C. R.; Richards, A. F. Indium(III) one-dimensional polymers with pyrazine and pyrazine, 2-carboxylic acid as the spacer molecules. Polyhedron 2007, 26 (4), 923-928.
  3. Wang, H.; Xu, X.; Li, X.; Zhang, J.; Li, C. Synthesis and sintering of indium tin oxide nanoparticles by citrate-nitrate combustion method. Rare Met. 2010, 29 (4), 355-360.