1-Naphthyl phosphate monosodium salt monohydrate, 98+%
1-Naphthyl phosphate monosodium salt monohydrate, 98+%
1-Naphthyl phosphate monosodium salt monohydrate, 98+%
1-Naphthyl phosphate monosodium salt monohydrate, 98+%
Thermo Scientific Chemicals

1-Naphthyl phosphate monosodium salt monohydrate, 98+%

CAS: 81012-89-7 | C10H7O4P | 222.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A18254.14
also known as A18254-14
Price (USD)
307.65
Online Exclusive
342.00
Save 34.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
307.65
Online Exclusive
342.00
Save 34.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS81012-89-7
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)3.53-9.89% (0.5-1.5 waters)
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
1-Naphthyl phosphate monosodium salt monohydrate acts as a substrate for determination of prostatic acid phosphatase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl phosphate monosodium salt monohydrate acts as a substrate for determination of prostatic acid phosphatase.

Solubility
Soluble in water.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Substrate for phosphatase: J. Biol. Chem., 7, 190 (1951).
  2. Renglin, A.; Olsson, A.; Wachtmeister, C. A.; Onfelt, A. Mitotic disturbance by carbaryl and the metabolite 1-naphthol may involve kinase-mediated phosphorylation of 1-naphthol to the protein phosphatase inhibitor 1-naphthylphosphate. Mutagenesis. 1998, 13 (4), 345-352.
  3. Bowers, J. L.; Bowers Jr, G. N. 1-naphthyl phosphate: specifications for high-quality substrate for measuring prostatic acid phosphatase activity. Clin. Chem. 1982, 28 (1), 212-215.