4-Nitroveratrole, 98+%
4-Nitroveratrole, 98+%
4-Nitroveratrole, 98+%
4-Nitroveratrole, 98+%
Thermo Scientific Chemicals

4-Nitroveratrole, 98+%

CAS: 709-09-1 | C8H9NO4 | 183.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
250 g
Catalog number A18264.18
also known as A18264-18
Price (USD)
73.60
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
73.60
Each
Add to cart
Chemical Identifiers
CAS709-09-1
IUPAC Name1,2-dimethoxy-4-nitrobenzene
Molecular FormulaC8H9NO4
InChI KeyYFWBUVZWCBFSQN-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1OC)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)94.5-100.5?C
FormCrystals or powder or crystalline powder
Appearance (Color)Yellow
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only