Calcium 2-ethylhexanoate, 98%
Calcium 2-ethylhexanoate, 98%
Calcium 2-ethylhexanoate, 98%
Thermo Scientific Chemicals

Calcium 2-ethylhexanoate, 98%

CAS: 136-51-6 | C16H30CaO4 | 326.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A18266.22
also known as A18266-22
Price (USD)
479.65
Online Exclusive
533.00
Save 53.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
479.65
Online Exclusive
533.00
Save 53.35 (10%)
Each
Chemical Identifiers
CAS136-51-6
IUPAC Namecalcium bis(2-ethylhexanoate)
Molecular FormulaC16H30CaO4
InChI KeyLTPCXXMGKDQPAO-UHFFFAOYNA-L
SMILES[Ca++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Calcium 2-ethylhexanoate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium 2-ethylhexanoate is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. E.Tkalcec; M.Sauer; R.Nonninger; H.Schmidt. Sol-gel-derived hydroxyapatite powders and coatings. Journal of Materials Science. 2001, 36,(21), 5253-5263.
  2. Hiroshi Kominami; Masaaki Kohnoa; Yoshiya Kera. Synthesis of brookite-type titanium oxide nano-crystals in organic media. J. Mater. Chem. 2000, 10,(5), 1151-1156.