Phenyl propargyl ether, 98+%
Phenyl propargyl ether, 98+%
Phenyl propargyl ether, 98+%
Thermo Scientific Chemicals

Phenyl propargyl ether, 98+%

CAS: 13610-02-1 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A18272.06
also known as A18272-06
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
114.00
Save 17.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
114.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS13610-02-1
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5315-1.5355 @ 20?C
Appearance (Color)Clear colorless to yellow to orange
Assay (GC)≥98.0%
FormLiquid
Phenyl propargyl ether is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl propargyl ether is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water (1.0 g/L at 25°C).

Notes
Air and light sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

General References:

  1. Walter S.Trahanovsky; Patrick W.Mullen. Formation of 2-indanone and benzocyclobutene from the pyrolysis of phenyl propargyl ether. J. Am. Chem. Soc. 1972, 94,(16), 5911-5913.
  2. M.V.Musalov; V.A.Potapov; S.V.Amosova. Annulation of phenyl propargyl ether with selenium dichloride. Russian Chemical Bulletin. 2011, 60,(4), 767-768.