5-Chlorobenzo-2,1,3-thiadiazole, 98%
5-Chlorobenzo-2,1,3-thiadiazole, 98%
5-Chlorobenzo-2,1,3-thiadiazole, 98%
Thermo Scientific Chemicals

5-Chlorobenzo-2,1,3-thiadiazole, 98%

CAS: 2207-32-1 | C6H3ClN2S | 170.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A18286.03
also known as A18286-03
Price (USD)
89.65
Online exclusive
99.80
Save 10.15
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
89.65
Online exclusive
99.80
Save 10.15
Each
Chemical Identifiers
CAS2207-32-1
IUPAC Name5-chloro-2,1,3-benzothiadiazole
Molecular FormulaC6H3ClN2S
InChI KeyVRNJWKISMWDTAY-UHFFFAOYSA-N
SMILESClC1=CC2=NSN=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)53.0-59.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only