4'-Benzyloxyacetophenone, 98%
4'-Benzyloxyacetophenone, 98%
4'-Benzyloxyacetophenone, 98%
4'-Benzyloxyacetophenone, 98%
Thermo Scientific Chemicals

4'-Benzyloxyacetophenone, 98%

CAS: 54696-05-8 | C15H14O2 | 226.275 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18302.14
also known as A18302-14
Price (USD)
219.65
Online Exclusive
244.00
Save 24.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
219.65
Online Exclusive
244.00
Save 24.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS54696-05-8
IUPAC Name1-[4-(benzyloxy)phenyl]ethan-1-one
Molecular FormulaC15H14O2
InChI KeyMKYMYZJJFMPDOA-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Melting Point (clear melt)90.5-96.5?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only