2-Methyl-2-butanol, 98%
2-Methyl-2-butanol, 98%
2-Methyl-2-butanol, 98%
Thermo Scientific Chemicals

2-Methyl-2-butanol, 98%

CAS: 75-85-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
500 mL
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A18304.AP
also known as A18304-AP
Price (USD)
65.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.60
Save 11.95
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.60
Save 11.95
Each
Chemical Identifiers
CAS75-85-4
IUPAC Name2-methylbutan-2-ol
Molecular FormulaC5H12O
InChI KeyMSXVEPNJUHWQHW-UHFFFAOYSA-N
SMILESCCC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4025-1.4075 @ 20?C
Appearance (Color)Clear, colorless
Assay (GC)≥97.5%
Identification (FTIR)Conforms
2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.

Solubility
Miscible with alcohol, ether, benzene, chloroform, glycerol, oils and acetone.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Nikoshvili, L.; Shimanskaya, E.; Bykov, A.; Yuranov, I.; Kiwi-Minsker, L.; Sulman, E. Selective hydrogenation of 2-methyl-3-butyn-2-ol over Pd-nanoparticles stabilized in hypercrosslinked polystyrene: Solvent effect. Catal. Today 2015, 241, 179-188.
  2. Vernuccio, S.; Von Rohr, P. R.; Medlock, J. General Kinetic Modeling of the Selective Hydrogenation of 2-Methyl-3-butyn-2-ol over a Commercial Palladium-Based Catalyst. Ind. Eng. Chem. Res. 2015, 54 (46), 11543-11551.