1,4-Diaminobutane dihydrochloride, 99%
1,4-Diaminobutane dihydrochloride, 99%
1,4-Diaminobutane dihydrochloride, 99%
Thermo Scientific Chemicals

1,4-Diaminobutane dihydrochloride, 99%

CAS: 333-93-7 | C4H14Cl2N2 | 161.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A18312.22
also known as A18312-22
Price (USD)
207.65
Online Exclusive
231.00
Save 23.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
207.65
Online Exclusive
231.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS333-93-7
IUPAC Namedihydrogen butane-1,4-diamine dichloride
Molecular FormulaC4H14Cl2N2
InChI KeyXXWCODXIQWIHQN-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)≤1%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only