2-Fluorophenyl isothiocyanate, 97%
2-Fluorophenyl isothiocyanate, 97%
2-Fluorophenyl isothiocyanate, 97%
Thermo Scientific Chemicals

2-Fluorophenyl isothiocyanate, 97%

CAS: 38985-64-7 | C7H4FNS | 153.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18314.06
also known as A18314-06
Price (USD)
42.65
Online Exclusive
47.10
Save 4.45 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
42.65
Online Exclusive
47.10
Save 4.45 (9%)
Each
Chemical Identifiers
CAS38985-64-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow or orange
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.6230-1.6280 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only