Manganese(II) bromide, anhydrous, 95%
Manganese(II) bromide, anhydrous, 95%
Manganese(II) bromide, anhydrous, 95%
Thermo Scientific Chemicals

Manganese(II) bromide, anhydrous, 95%

CAS: 13446-03-2 | MnBr2
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A18321.22
also known as A18321-22
Price (USD)
264.65
Special offer
Online exclusive
Ends: 30-Jun-2026
311.00
Save 46.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
264.65
Special offer
Online exclusive
Ends: 30-Jun-2026
311.00
Save 46.35 (15%)
Each
Chemical Identifiers
CAS13446-03-2
IUPAC Namemanganese(2+) dibromide
Molecular FormulaBr2Mn
InChI KeyRJYMRRJVDRJMJW-UHFFFAOYSA-L
SMILES[Mn++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink
FormPowder
Complexometric Titration≥94.0 to ≤106.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only