2-Bromobenzamide, 98%
2-Bromobenzamide, 98%
2-Bromobenzamide, 98%
Thermo Scientific Chemicals

2-Bromobenzamide, 98%

CAS: 4001-73-4 | C7H6BrNO | 200.035 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18331.06
also known as A18331-06
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.60
Save 9.95 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.60
Save 9.95 (14%)
Each
Chemical Identifiers
CAS4001-73-4
IUPAC Name2-bromobenzamide
Molecular FormulaC7H6BrNO
InChI KeyNHNAEZDWNCRWRW-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)156.0-163.0?C
Appearance (Color)White
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
2-Bromobenzamide has been used in microwave assisted one-pot synthesis of substituted 3-(phenylmethylene)isoindolin-1-ones, palladium-catalyzed synthesis of phenanthridinones and in the synthesis of new water-soluble iminophosphorane ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzamide has been used in microwave assisted one-pot synthesis of substituted 3-(phenylmethylene)isoindolin-1-ones, palladium-catalyzed synthesis of phenanthridinones and in the synthesis of new water-soluble iminophosphorane ligand.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Hellal M and Cuny GD. Microwave assisted copper-free Sonogashira coupling/5-exo-dig cycloisomerization domino reaction: access to 3-(phenylmethylene) isoindolin-1-ones and related heterocycles.Tetrahedron Lett.2011,52(42), 5508-5511.
  2. Chun Lu.; Anton V Dubrovskiy.; Richard C Larock. Palladium-catalyzed annulation of arynes by o-halobenzamides: synthesis of phenanthridinones.J Org Chem.2012,77(19), 8648-8656.