Diphenyl selenide, 97%
Diphenyl selenide, 97%
Diphenyl selenide, 97%
Thermo Scientific Chemicals

Diphenyl selenide, 97%

CAS: 1132-39-4 | C12H10Se | 233.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A18364.03
also known as A18364-03
Price (USD)
35.20
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
35.20
Each
Chemical Identifiers
CAS1132-39-4
IUPAC Name(phenylselanyl)benzene
Molecular FormulaC12H10Se
InChI KeyORQWTLCYLDRDHK-UHFFFAOYSA-N
SMILES[Se](C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6470-1.6510 @ 20°C
Appearance (Color)Clear pale yellow to orange or red to brown
FormLiquid
Assay (GC)≥96.0%
Diphenyl selenide is used for anti-selective Michael addition of thiols and their analogs to nitro-olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl selenide is used for anti-selective Michael addition of thiols and their analogs to nitro-olefins.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Mohan, B.; Yoon, C.; Jang, S.; Park, K. H. Copper Nanoparticles Catalyzed Se(Te)-Se(Te) Bond Activation: A Straightforward Route Towards Unsymmetrical Organochalcogenides from Boronic Acids. ChemCatChem 2015, 7 (3), 405-412.
  2. Guo, J.; Wen, Z.; Wang, Q.; Jin, J.; Ma, G. A conductive selenized polyacrylonitrile cathode material for re-chargeable lithium batteries with long cycle life. J. Mater. Chem. A 2015, 3 (39), 19815-19821.