4,4'-(Hexafluoroisopropylidene)diphenol, 98%
4,4'-(Hexafluoroisopropylidene)diphenol, 98%
4,4'-(Hexafluoroisopropylidene)diphenol, 98%
Thermo Scientific Chemicals

4,4'-(Hexafluoroisopropylidene)diphenol, 98%

CAS: 1478-61-1 | C15H10F6O2 | 336.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A18370.14
also known as A18370-14
Price (USD)
141.65
Special offer
Online exclusive
Ends: 30-Jun-2026
166.00
Save 24.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
141.65
Special offer
Online exclusive
Ends: 30-Jun-2026
166.00
Save 24.35 (15%)
Each
Chemical Identifiers
CAS1478-61-1
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)158-164?C
Appearance (Color)White to cream
FormPowder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
4,4'-(Hexafluoroisopropylidene)diphenol is used to cross-link and cure fluoroelastomers. is also used as a monomer in the synthesis of specialty polymers. Polymers containing Bisphenol AF are useful in high temperature composites, in electronic materials and other specialty applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-(Hexafluoroisopropylidene)diphenol is used to cross-link and cure fluoroelastomers. is also used as a monomer in the synthesis of specialty polymers. Polymers containing Bisphenol AF are useful in high temperature composites, in electronic materials and other specialty applications.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. M. Sava.; C. Chiriac.; C. Gaina.; A. Stoleriu.; V. Gaina. Synthesis and Characterization of Poly(Maleimide-Ethers) from the Reaction of Bisdichloromaleimides with 4,4'-(Hexafluoroisopropylidene)diphenol. Journal of Macromolecular Science, Part A: Pure and Applied Chemistry. 1997, 34 (4), 725-733.
  2. Peixiang Xing.; Gilles P. Robertson.; Michael D. Guiver.; Serguei D. Mikhailenko.; Serge Kaliaguine. Synthesis and characterization of poly(aryl ether ketone) copolymers containing (hexafluoroisopropylidene)-diphenol moiety as proton exchange membrane materials. Polymer. 2005, 46 (10), 3257-3263.