1-Fluoronaphthalene, 98%
1-Fluoronaphthalene, 98%
1-Fluoronaphthalene, 98%
Thermo Scientific Chemicals

1-Fluoronaphthalene, 98%

CAS: 321-38-0 | C10H7F | 146.164 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A18378.14
also known as A18378-14
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS321-38-0
IUPAC Name1-fluoronaphthalene
Molecular FormulaC10H7F
InChI KeyCWLKTJOTWITYSI-UHFFFAOYSA-N
SMILESFC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Refractive Index1.5910-1.5950 @ 20?C
Assay (GC)≥97.5%
1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Wojciech A. Majewski.; David F. Plusquellic.; David W. Pratt. The rotationally resolved fluorescence excitation spectrum of 1-fluoronaphthalene. J. Chem. Phys. 1989, 90 (3), 1362.
  2. J. Mispelter.; J.-Ph. Grivet.; J.-M. Lhoste. Electron spin resonance of phosphorescent fluorine-substituted aromatics I. 1-Fluoronaphthalene and 2-fluoronaphthalene in durene single crystals. Molecular Physics: An International Journal at the Interface Between Chemistry and Physics. 1971, 21 (6), 999-1013.