Erbium(III) oxalate decahydrate, 99.9%
Erbium(III) oxalate decahydrate, 99.9%
Erbium(III) oxalate decahydrate, 99.9%
Erbium(III) oxalate decahydrate, 99.9%
Thermo Scientific Chemicals

Erbium(III) oxalate decahydrate, 99.9%

CAS: 30618-31-6 | C6H26Er2O22 | 784.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A18420.22
also known as A18420-22
Price (USD)
270.65
Online Exclusive
300.00
Save 29.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
270.65
Online Exclusive
300.00
Save 29.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS30618-31-6
IUPAC Nametris(oxalic acid) dierbium decahydrate
Molecular FormulaC6H26Er2O22
InChI KeyCYHKKMLSGVLNPD-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.O.[Er].[Er].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofA≥99.85%
Appearance (Color)Pink
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only