2,3-Dichloro-1-propanol, 97+%
2,3-Dichloro-1-propanol, 97+%
2,3-Dichloro-1-propanol, 97+%
Thermo Scientific Chemicals

2,3-Dichloro-1-propanol, 97+%

CAS: 616-23-9 | C3H6Cl2O | 128.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18444.06
also known as A18444-06
Price (USD)
45.65
Online Exclusive
50.50
Save 4.85 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
45.65
Online Exclusive
50.50
Save 4.85 (10%)
Each
Chemical Identifiers
CAS616-23-9
IUPAC Name2,3-dichloropropan-1-ol
Molecular FormulaC3H6Cl2O
InChI KeyZXCYIJGIGSDJQQ-UHFFFAOYNA-N
SMILESOCC(Cl)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.0%
Refractive Index1.4825-1.4875 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only