1,5-Dibromopentane, 98%
1,5-Dibromopentane, 98%
1,5-Dibromopentane, 98%
Thermo Scientific Chemicals

1,5-Dibromopentane, 98%

CAS: 111-24-0 | C5H10Br2 | 229.943 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A18465.0E
also known as A18465-0E
Price (USD)
570.65
Special offer
Online exclusive
Ends: 30-Jun-2026
671.00
Save 100.35 (15%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
570.65
Special offer
Online exclusive
Ends: 30-Jun-2026
671.00
Save 100.35 (15%)
Each
Chemical Identifiers
CAS111-24-0
IUPAC Name1,5-dibromopentane
Molecular FormulaC5H10Br2
InChI KeyIBODDUNKEPPBKW-UHFFFAOYSA-N
SMILESBrCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5090-1.5145 @ 20?C
Assay (GC)≥97.5%
FormLiquid
1,5-Dibromopentane is used in the preparation of 1,5-di-Grignard reagent and novel spirocyclic pyrrolidones. It acts as a growth supplement for Yarrowia lipolytica 3589, a tropical marine yeast. It is involved in the synthesis of N-alkylated piperidine as well as tetrydrothiopyran (thiane) by reaction with primary amine and sodium sulfide respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dibromopentane is used in the preparation of 1,5-di-Grignard reagent and novel spirocyclic pyrrolidones. It acts as a growth supplement for Yarrowia lipolytica 3589, a tropical marine yeast. It is involved in the synthesis of N-alkylated piperidine as well as tetrydrothiopyran (thiane) by reaction with primary amine and sodium sulfide respectively.

Solubility
Miscible with water, slightly miscible with chloroform and benzene.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Lifa, T.; Tieu, W.; Hocking, R. K.; Codd, R. Forward and Reverse (Retro) Iron(III) or Gallium(III) Desferrioxamine E and Ring-Expanded Analogues Prepared Using Metal-Templated Synthesis from endo-Hydroxamic Acid Monomers. Inorg. Chem. 2015, 54 (7), 3573-3583.
  2. Perroni, D. V.; Baez-Cotto, C. M.; Sorenson, G. P.; Mahanthappa, M. K. Linker Length-Dependent Control of Gemini Surfactant Aqueous Lyotropic Gyroid Phase Stability. J. Phys. Chem. Lett. 2015, 6 (6), 993-998.