Manganese(II) nitrate tetrahydrate, 98%
Manganese(II) nitrate tetrahydrate, 98%
Manganese(II) nitrate tetrahydrate, 98%
Manganese(II) nitrate tetrahydrate, 98%
Thermo Scientific Chemicals

Manganese(II) nitrate tetrahydrate, 98%

CAS: 20694-39-7 | H8MnN2O10 | 251.006 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A18521.0B
also known as A18521-0B
Price (USD)
174.65
Special offer
Online exclusive
Ends: 28-Dec-2026
205.00
Save 30.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
174.65
Special offer
Online exclusive
Ends: 28-Dec-2026
205.00
Save 30.35
Each
Chemical Identifiers
CAS20694-39-7
IUPAC Namemanganese(2+) tetrahydrate dinitrate
Molecular FormulaH8MnN2O10
InChI KeyALIMWUQMDCBYFM-UHFFFAOYSA-N
SMILESO.O.O.O.[Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink
Trace MetalZinc (Zn) : ≤ 0.01%
Complexometric Titration≥97.5 to ≤102.5%
Chloride content≤ 0.005%
FormCrystalline powder or fused solid or lumps or chunks
View more
Manganese(II) nitrate tetrahydrate used as porcelain colorants. It is also used as a precursor for manganese dioxide and other manganese compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) nitrate tetrahydrate used as porcelain colorants. It is also used as a precursor for manganese dioxide and other manganese compounds.

Solubility
Soluble in ethanol and water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong reducing agents and strong acids.
RUO – Research Use Only

General References:

  1. Nkosi, F. P.; Jafta, C. J.; Kebede, M.; le Roux, L.; Mathe, M. K.; Ozoemena, K. I. Microwave-assisted optimization of the manganese redox states for enhanced capacity and capacity retention of LiAlxMn2- xO4 (x= 0 and 0.3) spinel materials. RSC Adv. 2015, 5 (41), 32256-32262.
  2. Baykal, A.; Güner, S.; Demir, A. Synthesis and magneto-optical properties of triethylene glycol stabilized Mn1-xZnxFe2O4 nanoparticles. J. Alloys Compd. 2015, 619, 5-11.