6-Bromo-2-naphthol, 97%
6-Bromo-2-naphthol, 97%
6-Bromo-2-naphthol, 97%
6-Bromo-2-naphthol, 97%
Thermo Scientific Chemicals

6-Bromo-2-naphthol, 97%

CAS: 15231-91-1 | C10H7BrO | 223.069 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A18535.18
also known as A18535-18
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS15231-91-1
IUPAC Name6-bromonaphthalen-2-ol
Molecular FormulaC10H7BrO
InChI KeyYLDFTMJPQJXGSS-UHFFFAOYSA-N
SMILESOC1=CC=C2C=C(Br)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pink to pale brown or grey
FormPowder
Assay (Silylated GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)125-132?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only