beta-Naphthoflavone, 98+%
beta-Naphthoflavone, 98+%
beta-Naphthoflavone, 98+%
Thermo Scientific Chemicals

beta-Naphthoflavone, 98+%

CAS: 6051-87-2 | C19H12O2 | 272.303 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A18543.06
also known as A18543-06
Price (USD)
114.65
Special offer
Online exclusive
Ends: 30-Jun-2026
135.00
Save 20.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
114.65
Special offer
Online exclusive
Ends: 30-Jun-2026
135.00
Save 20.35 (15%)
Each
Chemical Identifiers
CAS6051-87-2
IUPAC Name3-phenyl-1H-benzo[f]chromen-1-one
Molecular FormulaC19H12O2
InChI KeyOUGIDAPQYNCXRA-UHFFFAOYSA-N
SMILESO=C1C=C(OC2=C1C1=CC=CC=C1C=C2)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (HPLC)≥98.0%
Melting Point (clear melt)159.0-165.0?C
FormCrystals or powder or crystalline powder
Inhibitor of aryl hydrocarbon hydroxylase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only