2-Amino-6-bromobenzothiazole, 98%
2-Amino-6-bromobenzothiazole, 98%
2-Amino-6-bromobenzothiazole, 98%
Thermo Scientific Chemicals

2-Amino-6-bromobenzothiazole, 98%

CAS: 15864-32-1 | C7H5BrN2S | 229.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A18554.06
also known as A18554-06
Price (USD)
154.65
Special offer
Online exclusive
Ends: 30-Jun-2026
182.00
Save 27.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
154.65
Special offer
Online exclusive
Ends: 30-Jun-2026
182.00
Save 27.35 (15%)
Each
Chemical Identifiers
CAS15864-32-1
IUPAC Name6-bromo-1,3-benzothiazol-2-amine
Molecular FormulaC7H5BrN2S
InChI KeyVZEBSJIOUMDNLY-UHFFFAOYSA-N
SMILESNC1=NC2=CC=C(Br)C=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormPowder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only