Alginic acid sodium salt, very low viscosity
Alginic acid sodium salt, very low viscosity
Alginic acid sodium salt, very low viscosity
Thermo Scientific Chemicals

Alginic acid sodium salt, very low viscosity

CAS: 9005-38-3 | (C6H7O7)A(C6H7O7)BNa
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A18565.36
also known as A18565-36
Price (USD)
119.65
Special offer
Online exclusive
Ends: 30-Jun-2026
140.00
Save 20.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
Ends: 30-Jun-2026
140.00
Save 20.35 (15%)
Each
Chemical Identifiers
CAS9005-38-3
Molecular Formula(C6H7O7)A(C6H7O7)BNa
SMILES[Na+].O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C([O-])=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C([O-])=O
Molecular Weight (g/mol)NaN
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormPowder
Identification (FTIR)Conforms
Loss on Drying≤13.0%
pH5.0-7.5 (1% aq. solution)
View more
It is a hematologic agent, drug that acts on blood and blood-forming organs and those that affect the hemostatic system. sodium alginate has been used in warp sizing, packing and printing. In printing, it is an ideal kind of material with wide applications in the cotton, wool, silk and synthetic fiber. Sodium alginate is widely used in food industry such as it is the main material of seaweed, synthetic grape and synthetic cherry, it is taken as a stabilizer of ice cream and ice cream bar because of its dense organization and slow solution speed. It is also the main material of the dainty cold powder and jelly, As a stabilizer of cake (such as cookies, bread, fine dried noodle, chocolates) and a polish of bread, sodium alginate can keep the cookies sweet-smelling and avoid falling to pieces, and make the noodle smooth and reduce breaking ratio of noodle and fragmentation of bread. Sodium alginate is also a good thickener of jam, chili sauce, jelly, tomato ketchup, fishpaste, pudding and salad flavoring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only