Copper(II) formate tetrahydrate, 98%
Copper(II) formate tetrahydrate, 98%
Copper(II) formate tetrahydrate, 98%
Copper(II) formate tetrahydrate, 98%
Thermo Scientific Chemicals

Copper(II) formate tetrahydrate, 98%

CAS: 5893-61-8 | C2H10CuO8 | 225.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A18569.18
also known as A18569-18
Price (USD)
54.60
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
54.60
Each
Add to cart
Chemical Identifiers
CAS5893-61-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue to green
FormCrystals or Crystalline powder
Assay from Supplier's CofA≥97.5%
It is employed as an antibacterial agent in the treatment of cellulose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an antibacterial agent in the treatment of cellulose.

Solubility
Soluble in water (125 g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Hanako Kobayashi and Taiichiro Haseda. The Magnetic Properties of Cupric Formate Tetrahydrate at Low Temperatures.J. Phys. Soc. Jpn.1963,18541-550.
  2. Takasuke Matsuo.; Yoshio Kume.; Hiroshi Suga.; SyûzÔ Seki. Heat capacities of copper(II) formate tetrahydrate and tetradeuterate: A comparative study of phase transitions in layer hydrate crystals.J. Phys. Chem. Solids,1976,37(5), 499-506.