1,2-Cyclohexanediol, cis + trans, 98%
1,2-Cyclohexanediol, cis + trans, 98%
1,2-Cyclohexanediol, cis + trans, 98%
1,2-Cyclohexanediol, cis + trans, 98%
Thermo Scientific Chemicals

1,2-Cyclohexanediol, cis + trans, 98%

CAS: 931-17-9 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A18572.09
also known as A18572-09
Price (USD)
60.90
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
60.90
Each
Add to cart
Chemical Identifiers
CAS931-17-9
IUPAC Namecyclohexane-1,2-diol
Molecular FormulaC6H12O2
InChI KeyPFURGBBHAOXLIO-UHFFFAOYNA-N
SMILESOC1CCCCC1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps
Assay (GC)≥94.0%
Identification (FTIR)Conforms
Melting Point (clear melt)72.0-79.0°C
Appearance (Color)White to pale cream to pale brown
1,2-Cyclohexanediol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Cyclohexanediol is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. H. Stewart Hendrickson.; James L. Reinertsen. Comparison of metal-binding properties of trans-1,2-cyclohexanediol diphosphate and deacylated phosphoinositides. Biochemistry. 1969, 8 (12), 4855-4858.
  2. Gaetan Caron.; Romas J. Kazlauskas. An optimized sequential kinetic resolution of trans-1,2-cyclohexanediol. J. Org. Chem.. 1991, 56 (26), 7251-7256.