5-Nitro-2-furaldehyde semicarbazone, 98+%
5-Nitro-2-furaldehyde semicarbazone, 98+%
5-Nitro-2-furaldehyde semicarbazone, 98+%
5-Nitro-2-furaldehyde semicarbazone, 98+%
Thermo Scientific Chemicals

5-Nitro-2-furaldehyde semicarbazone, 98+%

CAS: 59-87-0 | C6H6N4O4 | 198.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A18593.14
also known as A18593-14
Price (USD)
38.50
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
38.50
Each
Add to cart
Chemical Identifiers
CAS59-87-0
IUPAC Name[(E)-[(5-nitrofuran-2-yl)methylidene]amino]urea
Molecular FormulaC6H6N4O4
InChI KeyIAIWVQXQOWNYOU-FPYGCLRLSA-N
SMILESNC(=O)N\N=C\C1=CC=C(O1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay (HPLC)≥98.0%
FormPowder
Bactericidal compound used as an antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only