2-Coumaranone, 97%
2-Coumaranone, 97%
2-Coumaranone, 97%
Thermo Scientific Chemicals

2-Coumaranone, 97%

CAS: 553-86-6 | C8H6O2 | 134.134 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A18617.06
also known as A18617-06
Price (USD)
121.65
Online Exclusive
135.00
Save 13.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
121.65
Online Exclusive
135.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS553-86-6
IUPAC Name2,3-dihydro-1-benzofuran-2-one
Molecular FormulaC8H6O2
InChI KeyACZGCWSMSTYWDQ-UHFFFAOYSA-N
SMILESO=C1CC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)45.0-52.0°C
Appearance (Color)White to pale cream to cream to yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only