4-Methoxytrityl alcohol, 94%
4-Methoxytrityl alcohol, 94%
4-Methoxytrityl alcohol, 94%
Thermo Scientific Chemicals

4-Methoxytrityl alcohol, 94%

CAS: 847-83-6 | C20H18O2 | 290.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A18620.18
also known as A18620-18
Price (USD)
160.65
Special offer
Online exclusive
Ends: 28-Dec-2026
189.00
Save 28.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
160.65
Special offer
Online exclusive
Ends: 28-Dec-2026
189.00
Save 28.35
Each
Chemical Identifiers
CAS847-83-6
IUPAC Name(4-methoxyphenyl)diphenylmethanol
Molecular FormulaC20H18O2
InChI KeyWCRRRAKYYPJJMP-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)73.0-86.0?C
Appearance (Color)White
Assay (Silylated GC)≥92.0%
It is used as an intermediate for preparation of TT-0011.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate for preparation of TT-0011.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Jasenka Matulic-Adamic.; Mark Sanseverino.; Leonid Beigelman. An efficient synthesis of the ribozyme-folate conjugate.Tetrahedron Lett.2002,43(25), 4439-4441.
  2. C. Bleasdale.; W. Clegg.; S. B. Ellwood and B. T. Golding. Structures of the tetrafluoroborate salts of triphenylcarbenium ions with one and with two 4-methoxy substituents: stable carbocations.Acta Cryst.1991,C47550-553.