Triphenyl phosphite, 97%
Triphenyl phosphite, 97%
Triphenyl phosphite, 97%
Thermo Scientific Chemicals

Triphenyl phosphite, 97%

CAS: 101-02-0 | C18H15O3P | 310.289 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number A18662.0E
also known as A18662-0E
Price (USD)
153.65
Online Exclusive
170.00
Save 16.35 (10%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
170.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS101-02-0
IUPAC Nametriphenyl phosphite
Molecular FormulaC18H15O3P
InChI KeyHVLLSGMXQDNUAL-UHFFFAOYSA-N
SMILESO(P(OC1=CC=CC=C1)OC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5885-1.5935 @ 20?C
Used to convert alcohols to alkyl halides, as a peptide coupling agent, and, in combination with ozone, as a low-temperature source of singlet oxygen, and ligand for metal-catalyzed reactions. Also used as reagent in the preparation of mixed valent nickel diphosphine dithiolate bridged iron carbonyl phosphine complexes as models of [NiFe]-hydrogenase active site.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to convert alcohols to alkyl halides, as a peptide coupling agent, and, in combination with ozone, as a low-temperature source of singlet oxygen, and ligand for metal-catalyzed reactions. Also used as reagent in the preparation of mixed valent nickel diphosphine dithiolate bridged iron carbonyl phosphine complexes as models of [NiFe]-hydrogenase active site.

Solubility
Insoluble in water.

Notes
Air and Moisture Sensitive, store away form air and moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
RUO – Research Use Only

General References:

  1. John D Meeker; and Heather M Stapleton. House dust concentrations of organophosphate flame retardants in relation to hormone levels and semen quality parameters. Environmental Health Perspectives, 2010, 118 (3), 318-323.
  2. Ken-ichiro Murata; and Hajime Tanaka. Surface-wetting effects on the liquid-liquid transition of a single-component molecular liquid. Nature Communications, 2010, 116.