CD4 Antibody, PE-Cy7® conjugate (RPA-T4)

Catalog number: A18706

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CD4 Monoclonal Antibody for Flow
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Human
Validated Application: Flow Cytometry
Host Species: Mouse
SubClass (Isotype): IgG1, kappa
Clonality: Monoclonal
Clone ID: RPA-T4
Antibody Fragment: Whole Antibody
Target Gene: CD4 (RPA-T4)
Gene ID: 920
Gene Symbol: CD4
Gene Aliases: CD4mut
Label or Dye: PE-Cy®7
Excitation⁄Emission (nm): 488⁄767
Excitation Class: Visible
Concentration: 0.1 mg⁄ml
Form: Liquid
Number of Reactions: 25 Tests
For Use With (Equipment): Attune® Acoustic Focusing Cytometer
Product Size: 25 tests
Other Additives: Sodium Azide Preservative

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products