Triethyl borate, 97%
Triethyl borate, 97%
Triethyl borate, 97%
Thermo Scientific Chemicals

Triethyl borate, 97%

CAS: 150-46-9 | C6H15BO3 | 145.993 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number A18728.0F
also known as A18728-0F
Price (USD)
578.00
Each
Quantity:
2500 mL
Request bulk or custom format
Price (USD)
578.00
Each
Chemical Identifiers
CAS150-46-9
IUPAC Nametriethyl borate
Molecular FormulaC6H15BO3
InChI KeyAJSTXXYNEIHPMD-UHFFFAOYSA-N
SMILESCCOB(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.3705-1.3775 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Triethyl borate is used as a solvent in resins, paints, varnishes, adhesives and sealant chemicals. It is used as a catalyst of synthetic waxes and resins. It is an active component of flame retardants in the textile industry and in welding fluxes. It is used to prepare m-tolyl-acetic acid ethyl ester by reacting with 1-bromomethyl-3-methyl-benzene and carbon monoxide using as1,5-hexadiene?dium chloride as a catalyst. Further, it is used in pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl borate is used as a solvent in resins, paints, varnishes, adhesives and sealant chemicals. It is used as a catalyst of synthetic waxes and resins. It is an active component of flame retardants in the textile industry and in welding fluxes. It is used to prepare m-tolyl-acetic acid ethyl ester by reacting with 1-bromomethyl-3-methyl-benzene and carbon monoxide using as1,5-hexadieneρdium chloride as a catalyst. Further, it is used in pyrotechnics.

Solubility
Miscible with alcohol and ether.

Notes
Moisture sensitive. Incompatible with water, strong oxidizing agents, mineral acids and strong acids.
RUO – Research Use Only

General References:

  1. Kim, M.; Jeon, S. K.; Hwang, S. H.; Lee, J. Y. Molecular design of triazine and carbazole based host materials for blue phosphorescent organic emitting diodes. Phys. Chem. Chem. Phys. 2015, 17 (20), 13553-13558.
  2. Kang, H.; Lee, H.; Shin, H.; Kang, S.; Kim, B.; Park, J. New Emitting Materials Based on HTL Moiety with High Hole Mobility for OLEDs. Mol. Cryst. Liq. Cryst. 2015, 618 (1), 47-54.