Ethyl 3-(2-furyl)propionate, 98%
Ethyl 3-(2-furyl)propionate, 98%
Ethyl 3-(2-furyl)propionate, 98%
Thermo Scientific Chemicals

Ethyl 3-(2-furyl)propionate, 98%

CAS: 10031-90-0 | C9H12O3 | 168.192 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18732.06
also known as A18732-06
Price (USD)
42.20
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
42.20
Each
Chemical Identifiers
CAS10031-90-0
IUPAC Nameethyl 3-(furan-2-yl)propanoate
Molecular FormulaC9H12O3
InChI KeyOWIWZQQFSTZZIG-UHFFFAOYSA-N
SMILESCCOC(=O)CCC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4580-1.4620 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only