3,4-Dinitrobenzoic acid, 98+%
3,4-Dinitrobenzoic acid, 98+%
3,4-Dinitrobenzoic acid, 98+%
3,4-Dinitrobenzoic acid, 98+%
Thermo Scientific Chemicals

3,4-Dinitrobenzoic acid, 98+%

CAS: 528-45-0 | C7H4N2O6 | 212.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A18774.14
also known as A18774-14
Price (USD)
244.65
Online Exclusive
272.00
Save 27.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
244.65
Online Exclusive
272.00
Save 27.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS528-45-0
IUPAC Name3,4-dinitrobenzoic acid
Molecular FormulaC7H4N2O6
InChI KeyOMVRRHJJQILIJX-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point161.5-167.5°C
Appearance (Color)Cream or pale yellow to yellow
Assay (Silylated GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only