Nerolidol, cis + trans, 97+%
Nerolidol, cis + trans, 97+%
Nerolidol, cis + trans, 97+%
Thermo Scientific Chemicals

Nerolidol, cis + trans, 97+%

CAS: 7212-44-4 | C15H26O | 222.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A18812.14
also known as A18812-14
Price (USD)
39.65
Online Exclusive
44.20
Save 4.55 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
44.20
Save 4.55 (10%)
Each
Chemical Identifiers
CAS7212-44-4
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.0% (sum of isomers)
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4770-1.4820 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only